missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STI1 Antibody (3E4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26299-50ul
This item is not returnable.
View return policy
Description
STI1 Monoclonal antibody specifically detects STI1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| STI1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 3E4 | |
| Western Blot 1:500-1:5000, ELISA | |
| Hop, HOPSTI1L, Hsc70/Hsp90-organizing protein, Hsp70/Hsp90-organizing protein, IEF-SSP-3521, NY-REN-11 antigen, Renal carcinoma antigen NY-REN-11, STI1P60, stress-induced-phosphoprotein 1, Transformation-sensitive protein IEF SSP 3521 | |
| A synthesized peptide derived from Human STI1 [UniProt P31948] | |
| 50 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10963 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction