missing translation for 'onlineSavingsMsg'
Learn More

STF-118804, MedChemExpress

Product Code. 16498077
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498077 5 mg 5mg
16485986 100 mg 100mg
16441557 50 mg 50mg
16425368 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498077 Supplier Medchem Express Supplier No. HY128085mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress STF-118804 is a highly specific NAMPT inhibitor; reduces the viability of most B-ALL cell lines with IC50 <10 nM.

STF-118804 is a highly specific NAMPT inhibitor

Tekniske data

Chemical Name or Material STF-118804
CAS 894187-61-2
Color White
Purity Grade Notes Research
Molecular Formula C25H23N3O4S
Quantity 5 mg
Solubility Information DMSO : ≥ 31 mg/mL (67.17 mM)
SMILES O=C(NCC1=CC=CN=C1)C2=CC=C(C3=NC(CS(=O)(C4=CC=C(C)C=C4)=O)=C(C)O3)C=C2
Molecular Weight (g/mol) 461.53
Formula Weight 461.53
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.