missing translation for 'onlineSavingsMsg'
Learn More

STF-083010, MedChemExpress

Product Code. 16456868
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456868 10 mg 10mg
16449407 5 mg 5mg
16441206 200 mg 200mg
16432937 50 mg 50mg
16426966 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16456868 Supplier Medchem Express Supplier No. HY15845
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress STF-083010 is a specific IRE1α inhibitor. STF-083010 inhibits Ire1 endonuclease activity, without affecting its kinase activity, after endoplasmic reticulum stress.

STF-083010 is a specific IRE1α inhibitor. STF-083010 inhibits Ire1 endonuclease activity, without affecting its kinase activity, after endoplasmic reticulum stress.

Specifications

Chemical Name or Material STF-083010
CAS 307543-71-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H11NO3S2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (315.08 mM; Need ultrasonic)
SMILES O=S(C1=CC=CS1)(/N=C/C2=C3C=CC=CC3=CC=C2O)=O
Molecular Weight (g/mol) 317.38
Formula Weight 317.38
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.