missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sterol carrier protein 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38141-20ul
This item is not returnable.
View return policy
Description
Sterol carrier protein 2 Polyclonal antibody specifically detects Sterol carrier protein 2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Sterol carrier protein 2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp686C12188, DKFZp686D11188, NLTP, non-specific lipid-transfer protein, NSL-TP, Propanoyl-CoA C-acyltransferase, SCP-2, SCP-CHI, SCPX, SCP-X, sterol carrier protein 2EC 2.3.1.176, Sterol carrier protein X | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-322 of human Sterol carrier protein 2 (NP_001007099.1).,, Sequence:, MSSSPWEPATLRRVFVVGVGMTKFVKPGAENSRDYPDLAEEAGKKALADAQIPYSAVDQACVGYVFGVAECVLALGFEKMSKGSLGIKFSDRTIPTDKHVDLLINKYGLSAHPVAPQMFGYAGKEHMEKYGTKIEHFAKIGWKNHKHSVNNPYSQFQDEYSLDEVMASKEVFDFLTILQCCPTSDGAAAAILASEAFVQKYGLQSKAVEILAQEMMTDLPSSFEEKSIIKMVGFDMSKEAARKCYEKSGLTPNDIDVIELHDCFSTNELLTYEALGLCPEGQGATLVDRGDNTYGGKWVINPSGGLISKGHPLGATGGHSCS | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6342 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction