Learn More
Sampling Systems™ SteriWare™ Sterile HDPE Disposable Powder Thief
Sampling Systems™ SteriWare™ Sterile HDPE Disposable Powder Thief is ideal for taking representative samples from known locations within the bulk powder.
Brand:  Sampling Systems™ 8060H-505S
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11905636
  • £128.00 / Pack of 20
Estimated Shipment: 15-07-2024
to see stock.
Add to basket
Description
Description
  • Assembled and packed in a cleanroom
  • Individually Bagged
  • Made from FDA or EU conforming HDPE
  • Available as gamma irradiated sterile (SAL 10-6)
  • BSE/TSE Free
  • Full Batch Traceability
  • Suitable for use in ATEX zones 0, 1 and 2
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • No cleaning validation (this is very expensive to set up and run)
  • Manufactured in a class 100,000 cleanroom environment (check individual sampler descriptions)
  • Peace of mind
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Financial
  • Save time and money (it will cost more to clean a reusable sampler than to buy a disposable sampler)
Specifications
Specifications
HDPE
Sterile
500 mm
5 mL
21 mm