missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Slot Samplers
BIOPHARMA

Product Code. 17504606 Shop All Sampling Systems Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 20
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
17504606 Sterile Pack of 20
17594596 Non-sterile Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17504606 Supplier Sampling Systems™ Supplier No. 8073H300S
Add to basket
Add to basket
This item is not returnable. View return policy

Improve your powder sampling with the SteriWare™ Slot Sampler. With single use Slot Samplers you will be able to quickly take multilevel samples from a wide range of powders and granules. SteriWare™ Slot Samplers are widely used throughout the food and pharmaceutical industries.

  • The sterile, disposable Slot Sampler is ideal for taking a multilevel, cross sectional sample from powders and granules.
  • It is available in 2 diameters which gives the option of standard volume or large volume sampling from multiple points.
  • The standard 25 mm diameter version is available in various lengths.
  • Simple to use.
    • Push the sampler into the powder and rotate the handle to open the sampler and take the sample.
    • Close and remove the sampler.
  • Made from food grade HDPE.
  • Conforms to the latest FDA and EU requirements.
  • BSE/TSE free.
  • The samplers are manufactured, assembled and packed in a cleanroom to ensure cleanliness.

Specifications

Material HDPE
Capacity (Metric) 75 mL
Disposable Single-use
Sterility Sterile
Packaging Type Individually Bagged
Length (Metric) 420 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.