Learn More
Sampling Systems™ SteriWare™ Scraper
BIOPHARMA
Product Code. 15164966 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
190 mm
225 mm
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Sterility unitSize
15164966 - - -
13462829 225 mm Sterile Pack of 100
15879561 225 mm Non-sterile Pack of 100
15889561 190 mm Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15164966 Supplier Sampling Systems™ Supplier No. 8785H75S
  • ONLINE EXCLUSIVE PRICE
    £211.03
    £180.00 / Pack of 100
    Save £31.03
    14% Off
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
Designed to be ergonomic, which means they are easy to use and ideal for scraping and cleaning. Sampling Systems™ SteriWare™ Scraper is ideal for use with foodstuffs, cosmetics and pharmaceuticals.
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Available in 2 sizes
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contaminaton
  • No cleaning required