Learn More
Sampling Systems™ SteriWare™ Micro-Spatulas
BIOPHARMA
Product Code. 17534566 Shop All Sampling Systems Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
0.3 mL
0.5 mL
2 mL
Color:
Blue
Red
White
missing translation for 'unitSize'
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Capacity (Metric) Color unitSize
17534566 0.3 mL Red Pack of 100
17524566 0.3 mL Blue Pack of 100
17514566 0.5 mL Blue Pack of 100
17504566 2 mL White Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17534566 missing translation for 'mfr' Sampling Systems™ missing translation for 'supplierNo' 8015VR170S
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
SteriWare™ Micro-Spatulas are ideal for single use applications in pharmaceutical, chemical and other hygienic industries.
  • The disposable Micro Spatulas are ideal for a wide range of activities including sampling, dispensing and measuring small amounts of powder and granule.
  • The narrow blades give you total control as well as enabling use in small spaces such as vials.
  • The single-use spatula is ideal when costly cleaning and cleaning validation needs to be avoided.
  • Simply unwrap the sampler and use it.
  • It has been made in a controlled clean environment (class 100,000 cleanroom).
  • They are available in white, blue and red.
  • Colored versions are ideal if you are concerned about leaving it in the powder - they can be easily seen.
  • Used by pharmaceutical, food and chemical companies around the world.
  • Made from high-quality PS, PP or HDPE that conforms to the latest food grade, EC and FDA standards.
  • Sterility Assurance Level (SAL) of 10-6
  • BSE/TSE free.
  • Batch certificates of conformity supplied.
Spezifikation
Sterility Sterile
Capacity (Metric) 0.3 mL
Color Red
Disposable Single-use
Material Polystyrene
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.