Learn More
Buerkle™ SteriPlast™/LaboPlast™ Palette Knife Spatula
GREENER_CHOICEBIOPHARMA
Product Code. 17290584 Shop All Bürkle Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility unitSize
17290584 Sterile Pack of 100
17270584 Non-sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17290584 Supplier Buerkle™ Supplier No. 53781035
Only null left
Add to basket
Add to basket
Can be used to apply, distribute or evenly spread viscous media such as pastes and creams. The spatula reaches easily into the corners and curves of various containers and is suitable for scraping out bowls, removing residue or wiping off Bürkle's dosing spoons.
  • The dimensionally stable long edge of the plastic scraper is ideal for cutting viscous media.
  • A small tip on the long edge can be used for ripping, for example packaging film.
  • The scraper is also suitable for stirring and blending.
  • PS, white
  • Available either in a large package with 100 pieces or 100 pieces packaged individually and radiation sterilised
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100
Sterility Sterile
Autoclavable Not Autoclavable
Certifications/Compliance Complies with EU food rrequirements and FDA regulations
Disposable Single-use
Product Type Sampler
Length (Metric) Blade 70 mm
Material Polystyrene
Length (Metric) 192 mm