Learn More
Gibco™ Sterilized Luer Connectors compatible with CTS™ Rotea™ Single-Use Kit
Description
The Sterilized Luer Connectors, compatible with the CTS Rotea Single-Use kits, enable connection of bags without the need for a tube welder. Each pack of connectors is double bagged and gamma irradiated.
This product includes 10 packs of connectors. Each pack contains the following Luer connectors:
- 8 x female Leur to barb
- 8 x male Luer to barb
- 8 x female Luer cap
- 8 x male Luer plug
Specifications
Specifications
| Refrigerated | No |
| Product Line | CTS Rotea |
| Quantity | 10 x 32 Connectors |