missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Plastic Round Base Tubes

Product Code. 12852863
Change view
Click to view available options
Tubing Size:
0.6 mL
1.0 mL
2.5 mL
2.7 mL
4.9 mL
11.0 mL
Packaging Quantity:
250
500
1000
1500
2500
Unit Size:
Pack of 10000
Pack of 2500
Pack of 3500
Pack of 6000
Pack of 750
Product Code. Tubing Size Packaging Quantity unitSize
12852863 11.0 mL 1500 Pack of 750
11748868 0.6 mL 1000 Pack of 10000
13015723 1.0 mL 1000 Pack of 10000
12842863 2.5 mL 2500 Pack of 2500
11708988 2.7 mL 500 Pack of 6000
11778908 4.9 mL 250 Pack of 3500
6 options
This item is not returnable. View return policy

Product Code. 12852863

Brand: Thermo Scientific™ 30932

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene.

Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile.

Not available in all geographic regions.

  • Suitable for samples ranging from 0.6 to 11mL
  • Supplied nonsterile
TRUSTED_SUSTAINABILITY

Specifications

Description Round Base Plastic Tubes
Material Polystyrene
Diameter (Metric) Outer 16 mm
Packaging Quantity 1500
Product Line Sterilin
Sterility Non-sterile
Tubing Size 11.0 mL
Product Type Base Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.