Learn More
Thermo Scientific™ Sterilin™ Plastic Round Base Tubes
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene.
Brand: Thermo Scientific™ 30908
Code : 04
Additional Details : Weight : 4.66000kg
Description
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile. Not available in all geographic regions.
- Suitable for samples ranging from 0.6 to 11mL
- Supplied nonsterile
Specifications
Round Base Plastic Tubes | |
11 mm | |
Non-sterile |
Polystyrene | |
2500 | |
2.5 mL |