missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Plastic Round Base Tubes

Product Code. 11778908
Click to view available options
Tubing Size:
0.6 mL
1.0 mL
2.5 mL
2.7 mL
4.9 mL
11.0 mL
Packaging Quantity:
250
500
1000
1500
2500
Unit Size:
Pack of 10000
Pack of 2500
Pack of 3500
Pack of 6000
Pack of 750
This item is not returnable. View return policy

Product Code. 11778908

Brand: Thermo Scientific™ RT30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene.

Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile.

Not available in all geographic regions.

  • Suitable for samples ranging from 0.6 to 11mL
  • Supplied nonsterile
TRUSTED_SUSTAINABILITY

Specifications

Description Round Base Plastic Tubes
Material Polystyrene
Diameter (Metric) Outer 12 mm
Packaging Quantity 250
Product Line Sterilin
Sterility Non-sterile
Tubing Size 4.9 mL
Product Type Base Tube
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.