Learn More
Thermo Scientific™ Sterilin™ Plastic Round Base Tubes
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene.
Brand: Thermo Scientific™ RT25
Code : 56
Additional Details : Weight : 9.24000kg
Description
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic Round Base Tubes are manufactured from virgin polystyrene and supplied nonsterile. Not available in all geographic regions.
- Suitable for samples ranging from 0.6 to 11mL
- Supplied nonsterile
Specifications
Round Base Plastic Tubes | |
10 mm | |
Non-sterile |
Polystyrene | |
500 | |
2.7 mL |