Learn More
Thermo Scientific™ Sterilin™ Plain Swabs, General purpose
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport.
Brand: Thermo Scientific™ 155C
Description
Simplify sample collection with Thermo Scientific™ Sterilin™ Plain Swabs. Ideal for samples that will be processed immediately and don't require preservative for transport. Supplied in either a peel pouch or in a tube for ease of handling and transportation.
AVAILABLE IN UK ONLY
- Primary sample collection device for bacteria
- Color-coded caps for ease of identification
- CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
- Must be processed as soon as possible after sampling (ideally within two-hour period) to avoid dehydration of the sample and possible nonrecovery of pathogens
- Not available in all geographic regions
Specifications
| Irradiated | |
| CE Marked as Class IIa in accordance with the Medical Device Directive93/42/EEC (for transient invasive use) | |
| Tube | |
| 1 | |
| White |
| Plastic | |
| Rayon | |
| General purpose | |
| 100 |