Learn More
Thermo Scientific™ Sterilin™ Plain Flocked Swabs, Regular nylon flocked swab
Optimize your samples with our innovative nylon fiber technology flocked swabs. Thermo Scientific™ Sterilin™ Plain Flocked Swabs provide both outstanding collection and up to 95% release.
Brand:  Thermo Scientific™ 502CS01
Product Code. 11379173
  • £276.62 / Pack of 100
Edge
Description
Description
Optimize your samples with our innovative nylon fiber technology flocked swabs. Thermo Scientific™ Sterilin™ Plain Flocked Swabs provide outstanding collection. Sample release from the flocked swab is at least 95% compared with 25% with a regular fiber swab, which is critical if the sample to be collected is particularly small.
  • PCR compatible for molecular analysis
    • Manufactured from completely inert components
    • Certified RNase/DNase-free
    • For forensic purposes product is available certified human DNA free and with a molded breakpoint so that it can be easily broken into a 2mL collection vial
  • Available either in a paper peel pouch or in a plain dry tube
Certifications
  • CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
Ordering Information
  • Available in UK only
Specifications
Specifications
Regular nylon flocked swab
Plastic
Nylon
Individually wrapped in paper peel pouch
80mm
Irradiated
CE Marked as Class IIa in accordance with the Medical Device Directive93/42/EEC (for transient invasive use); Certified Human DNA-free, RNase/DNase-free
Regular Flocked
100
Flocked swabs - Individually wrapped in paper peel pouch