Learn More
Thermo Scientific™ Sterilin™ Gel Transport Swabs
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
Description
Ensure optimal sample collection and recovery using Thermo Scientific™ Sterilin™ Gel Transport Swabs. Swabs contain Amies media to preserve sample during lab transport.
AVAILABLE IN UK ONLY
- Contains media for the maintenance of bacterial sample during transport to the laboratory
- In vitro use only
- Each batch of product is tested for performance using a wide range of aerobic and anaerobic pathogens to ensure adequate recoveries
- Certificate of Sterility and Quality Assurance is available on request
- Lot number/expiry date and tamper-evident tube ensure complete traceability and sterility
Transport, M40
- Complies with American NCCLS M40-A and the German DIN 58942-4 standards
- Extended length of time for bacterial survival—48 hours (minimum)
- Wider temperature range—comparable performance at both room temperature and 4°C
Certifications
- CE marked as Class IIa in accordance with the Medical Device Directive 93/42/EEC (for transient invasive use)
- Not available in all geographic regions