missing translation for 'onlineSavingsMsg'
Learn More

Sterilin™ Colored Polystyrene Petri Dishes

Brand:  Thermo Scientific Sterilin 101VAMB

Additional Details : Weight : 9.08000kg

 View more versions of this product

Product Code. 11768948

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

EN ISO 24998:2008 standard
Polystyrene
Sterile
For In vitro use only
16.2 mm
Amber
Round
Special Petri Dish
500 Case
90 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers