Learn More
Thermo Scientific™ Sterilin™ Autoclave Bags, Coated wire holder for 511 bags
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
Brand: Thermo Scientific™ S23E
Code : 56
Additional Details : Weight : 0.21000kg
Description
Thermo Scientific™ Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
- HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
- Convenient “tissue box” cartons dispense bags individually
- Coated wire stands or cardboard holders available for use with autoclave bags
Ordering Alerts:
Not available in all geographic regions
Specifications
Coated wire holder for 509 and 510 bags | |
Coated Wire | |
1 |