Learn More
Bel-Art™ Sterileware™ Powder Thief
Bel-Art™ Sterileware™ Powder Thief is available in two lengths and four tip volumes and easily samples from targeted locations within bulk powders.
Brand: Bel-Art™ F37874-0005
Additional Details : Weight : 2.18300kg
Description
- Used by pharmaceutical, food, cosmetic and chemical companies.
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
- Gamma irradiated sterile (SAL 10-6)
- Ready for one time use and disposal
- Lot stamped for accurate tracing
- Shipped with a Certificate of Processing for Sterilization
Specifications
HDPE | |
Sterile | |
19 3/4 in. |
5 mL | |
20/Pk. | |
50 cm |