Learn More
Bel-Art™ Sterileware™ Powder Thief
Bel-Art™ Sterileware™ Powder Thief is available in two lengths and four tip volumes and easily samples from targeted locations within bulk powders.
Brand:  Bel-Art™ F37874-1010
Additional Details : Weight : 3.84990kg
 View more versions of this product
Product Code. 16632834
  • £256.00 / Pack of 20
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • Used by pharmaceutical, food, cosmetic and chemical companies.
  • Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
  • High density polyethylene (HDPE) thief heat sealed in individual polyethylene bag
  • Gamma irradiated sterile (SAL 10-6)
  • Ready for one time use and disposal
  • Lot stamped for accurate tracing
  • Shipped with a Certificate of Processing for Sterilization
Specifications
Specifications
HDPE
Sterile
39 3/8 in.
10 mL
20/Pk.
100 cm