missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Ensures cleanroom is free from contamination and particulates
- A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, moulds, yeasts, viruses and spores
- It is 0.2 micron filtered and filled in a Grade A environment
- Provided double bagged, the product is designed for ease of entry into pharmaceutical cleanrooms
- Log 6 kill against Bacillus subtilis in 60 sec. saves time spent on bio-decontamination
- Filtered to 0.2 microns in a Grade B cleanroom ensures the product is free from contamination and particulates
- Sterile version available for Grade A and B cleanrooms
- Contains no quaternary ammonium or surfactant so it's very low residue
- No hazard classification means only basic PPE required and no special disposal is required
- Double bagged packed in linear tear packaging so it's easy to open even when wearing gloves
Specifications
Specifications
| Product Type | Sporicide |
| Form | Liquid |
| Sterility | Sterile |
| Appearance | Colorless |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Disposable | Limited-use |
| Filtered | Filtered |
| Flammable | No |
| Quantity | 5 L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?