missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Technical Service Consultants™ TSC Neutralizing Buffer
Click to view available options
Quantity:
160 Each
Description
- Recommended for: hygiene surveillance (HACCP) and environmental monitoring of most surfaces where chemical residue neutralisation is recommended
- Ideal for whole sample analysis; food security from 'farm to fork' scenarios, including TACCP validation; suitable for clinical and veterinary infection control settings
- Excellent recovery of organisms including TVC, Listeria spp., Staphylococci, Enterobacteriaceae eg Salmonella spp, E.coli (E coli / ecoli / e. coli), Enterobacter, staphylococcus, klebsiella, E. coli Enterobacter, Enterobacter sakazakii
- Known limitations:
- Analyze as soon as possible due to the small volume to minimize sample dryout and organism loss
- Refrigeration will stabilize recovery of some organisms
- Not suitable for most fastidious organisms unless analyzed immediately
- Shelf life: 12 months
- Sterilised by gamma irradiation to SAL of 10-6
Specifications
Specifications
| Chemical Name or Material | Neutralizing buffer |
| Description | Polystyrene shaft |
| Quantity | 160 Each |
| Packaging | Hygiene Swab |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction