Learn More
Greiner Bio-One 384 Well Microplates
Product Code. 10497092
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
Product Code. Color unitSize
10497092 Clear Pack of 100
1 options
Product Code. 10497092 Supplier Greiner Bio-One Supplier No. 781162
Only null left
Add to basket
Add to basket
Designed with a rounded square profile for flexible working volume combined with reduced wicking and bubbling. Greiner Bio-One™ 384 Well Polystyrene Microplates provide four times the number of wells in a 96 well standard microplate while retaining external dimensions compatible with standard equipment and automated systems.
  • Clear polystyrene construction for optimum visibility
  • Footprint compatible with automated systems
  • Can be traced to production through defined LOT number system
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Well-to-well spacing: 4.5mm
  • Square wells with rounded corners combines advantages of square well (i.e. flexible working volume of 10μ to 130μ with the advantages of a round well (reduced wicking and bubbling) 
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN ISO 9001
Certified DNase/RNase and human DNA free
Format Standard
Array 16 x 24
Color Clear
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 10 to 130 μL
Sterility Sterile
Total Volume (Metric) Well 138 μL
Well Shape Flat Bottom (F-well)
No. per Pack 100Pack
Quantity 100/Pk.