missing translation for 'onlineSavingsMsg'
Learn More

AGMA Sterile Neutral Detergent

Product Code. p-200087503
Click to view available options
Container Type:
Capped Bottle
Trigger Spray
Quantity:
6 x 900 mL
20 x 500 mL
2 x 5 L
15 x 1 L
Unit Size:
Pack of 15
Pack of 2
Pack of 20
Pack of 6
This item is not returnable. View return policy

Product Code. 30066761

Brand: AGMA 0008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protect your cleanrooms with a highly-effective, deep cleaning Sterile Neutral detergent.

Implementing The considerable expertise in surfactant technology and disinfection, The Sterile Neutral Detergent is designed specifically for use in cleanrooms.

The Sterile Neutral Detergent formulation is available in a range of formats, including a 500 mL and 900 mL trigger spray, 1 L and 5 L fluid container or impregnated onto The high quality, low linting 68 gsm wipes or economy 4YPINs.

It is available as a gamma irradiated Sterile solution, ensuring product suitability no matter the class of your cleanroom.

With highly effective surface cleaning properties, it is ideal when a clean is required.

  • Formulated to be low foaming, it does not leave significant residues or smears
  • 0.2 μm filtered fluid and gamma irradiated guaranteeing sterility
  • Double bagged (where applicable)
  • Trigger sprays prepared under Grade A conditions in a Grade C/D cleanroom
  • Bag in bottle system ensures in-use sterility (Trigger Spray only)
  • 13 week weeks in-use validation (Trigger Spray only)
  • Wipes impregnated in a Grade C environment
  • Blended with WFI quality water to help ensure a low endotoxin level
  • 2 year unopened shelf life
  • Certification available for full traceability Available in a range of formats to suit your SOPs
  • 500 mL and 900 mL trigger sprays for ease of use by operators
  • Convenient 1 L or 5 L fluid for dosing dry wipes or use in mopping systems

Especificaciones

Product Type Neutral Detergent
Form Liquid
pH 6 to 9
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Composition Neutral Detergent in WFI
Container Type Capped Bottle
Disposable Limited-use
Filtered Filtered
Flammable No
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Quantity 2 x 5 L
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado