missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Sterile Inoculation Loop

Product Code. 10659021
Change view
Click to view available options
Color:
Blue
White
Yellow
Quantity:
2000/Pk.
600/Pk.
Unit Size:
Pack of 2000
Pack of 600
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
10659021 White 2000/Pk. Pack of 2000
10707682 Blue 600/Pk. Pack of 600
10342651 Yellow 600/Pk. Pack of 600
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10659021 Supplier Greiner Bio-One Supplier No. 731101
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Transfers inoculum for streaking. Greiner Bio-One™ Sterile Inoculation Loops are ideal for retrieving an inoculum from a culture of microorganisms.

  • Flexible loops for easier collection and inoculation
  • Sterilized by irradiation
  • For research only; not for use in diagnostic procedures, unless otherwise indicated
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Volume 1 μL
Length (Metric) 200 mm
Sterility Sterile
Color White
Quantity 2000/Pk.
For Use With (Application) Research use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.