missing translation for 'onlineSavingsMsg'
Learn More

Technical Service Consultants™ Sterile Inoculation Loop

Product Code. 12769875
Change view
Click to view available options
Volume:
1 μL
5 μL
10 μL
Color:
Clear
Green
Red
White
Yellow
Quantity:
1000 Pk.
20/bag
50 bags of 20
50/bag
Unit Size:
Pack of 1000
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume Color Quantity Unit Size
12769875 1 μL Yellow 20/bag Pack of 1000
12371309 10 μL Green 20/bag Pack of 1000
12789865 10 μL Green 50/bag Pack of 1000
12749865 10 μL White 50 bags of 20 Pack of 1000
12769865 10 μL White 1000 Pk. Pack of 1000
12381309 1 μL Clear 20/bag Pack of 1000
12709875 5 μL Red 20/bag Pack of 1000
12729875 5 μL Red 1000 Pk. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 12769875 Supplier Technical Service Consultants™ Supplier No. TS/4D
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use the Technical Service Consultants™ Sterile Inoculation Loop for general purpose use or routine samples. Its soft HI polystyrene composition prevents the head breaking off, which makes it ideal for clinical use.

  • Laboratory use only
  • Shelf Life: 3 years.
  • Sterilization Method: Gamma Irradiation to achieve a SAL of 10-6
  • 4-25°C. Store in dry and ventilated area; protect from light, heat and moisture in tightly closed original container

Specifications

Disposable Single-use
Volume 1 μL
Material Polystyrene
Sterility Sterile
Color Yellow
Product Type Inoculating Loop
For Use With (Equipment) Laboratory use only
Quantity 20/bag
For Use With (Application) Laboratory use only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.