missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gosselin™ Sterile Graduated Pipette
Used in dairy, environment, industries and microbiology applications.
Brand: Gosselin™ GPN1E25
Code : NEW
Additional Details : Weight : 12.82500kg
Description
- Used in dairy, environment, industrial and microbiological applications
Specifications
ISO 11137 : STERILE SAL 10-3 | |
Polystyrene | |
Yes | |
270 mm |
With yellow plug | |
Sterile | |
1 mL | |
Pipette |