missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Sterile Glass Serological Pipet

Graduated serological pipettes

Brand:  Poulten Graf™ D806/PS

Code : 02

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12344448

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Graduated glass pipettes
Specifications

Specifications

Glass
Yes
500/Pk.
260 mm
Single-use
Sterile
0.02 mL
2 mL
Serological Pipette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers