missing translation for 'onlineSavingsMsg'
Learn More

AGMA Sterile Alkaline Detergent

Product Code. 30066729
Click to view available options
Container Type:
Capped Bottle
Trigger Spray
Quantity:
6 x 900 mL
2 x 5 L
15 x 1 L
Unit Size:
Pack of 15
Pack of 2
Pack of 6
This item is not returnable. View return policy

Product Code. 30066729

Brand: AGMA 0096

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protect your cleanrooms with a highly-effective, deep cleaning Sterile Alkaline detergent

Sterile Alkaline Detergent effectively removes stubborn contaminants prior to disinfection, including visible soiling or build-up of residue from previous disinfectant applications.

With a pH of 12, it is highly effective at breaking down proteins, which makes it perfect for the first step in cleaning up spills or for preliminary cleans of new manufacturing cleanrooms.

  • A pH of 12is ideal for deep cleans and high protein load cleaning.
  • Particularly effective against proteinaceous and organic residues.
  • Removes build-up that neutral or acidic cleaners may not.
  • Low foaming
  • Does not leave significant residues.
  • Made with WFI water.
  • Manufactured in Class C/D cleanrooms.
  • Quality Management Certified to BS EN ISO 9001:2015.
  • Manufactured in accordance with the cGMP Guidelines.
  • Fully traceable with supporting analysis.
  • Available in 1 L and 5 L cap bottles, and 900 mL trigger bottles
  • Wipes impregnated in a Grade C environment
  • Blended with WFI quality water to help ensure a low endotoxin level
  • 2 year unopened shelf life
  • Certification available for full traceability Available in a range of formats to suit your SOPs
  • 500 mL and 900 mL trigger sprays for ease of use by operators
  • Convenient 1 L or 5 L fluid for dosing dry wipes or use in mopping systems

Specifications

Product Type Sterile Alkaline Detergent in WFI
Color-coded No
Form Liquid
pH 11.5 to 12.5
Scent Unscented
Sterility Sterile
Appearance Colorless
Autoclavable Not Autoclavable
Color Clear
Container Type Capped Bottle
Disposable Limited-use
Filtered Filtered
Flammable No
For Use With (Application) Cleanrooms and/or controlled environments subject to your contamination control strategies
Packaging Individually Double Bagged
Quantity 15 x 1 L
Show More Show Less
compliance-icons
Product Identifier
  • Sterile Alkaline Detergent
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : 2-(2-BUTOXYETHOXY)ETHANOL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.