missing translation for 'onlineSavingsMsg'
Learn More

Sterigmatocystin, TRC

Product Code. 30010488
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010488 1 mg 1mg
30006393 5 mg 5mg
30005609 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010488 Supplier Toronto Research Chemicals Supplier No. TRCS6867101MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10048-13-2
Molecular Formula C18 H12 O6
Molecular Weight (g/mol) 324.28
Synonym 7H-Furo[3',2':4,5]furo[2,3-c]xanthen-7-one, 3a,12c-dihydro-8-hydroxy-6-methoxy- (7CI,8CI), 7H-Furo[3',2':4,5]furo[2,3-c]xanthen-7-one, 3a,12c-dihydro-8-hydroxy-6-methoxy-, (3aR-cis)-, Sterigmatocystin (6CI), (3aR,12cS)-3a,12c-Dihydro-8-hydroxy-6-methoxy-7H-furo[3',2':4,5]furo[2,3-c]xanthen-7-one, NSC 201423, NSC 204985, Sterigmatocystine
SMILES COc1cc2OC3OC=CC3c2c4Oc5cccc(O)c5C(=O)c14

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H12O6/c1-21-11-7-12-13(8-5-6-22-18(8)24-12)17-15(11)16(20)14-9(19)3-2-4-10(14)23-17/h2-8,18-19H,1H3
Formula Weight 324.0634
Chemical Name or Material Sterigmatocystin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.