missing translation for 'onlineSavingsMsg'
Learn More

Merck Stericup™ Quick Release-HV Sterile Vacuum Bottle Top Filtration Systems

Product Code. 15908006 Shop All Merck Products
Change view
Click to view available options
Volume (Metric):
150 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Product Code. Volume (Metric) unitSize
15908006 250 mL Pack of 12
15997996 150 mL Pack of 12
15918006 500 mL Pack of 12
15928006 1000 mL Pack of 12
4 options
This item is not returnable. View return policy

Product Code. 15908006

Brand: Merck S2HVU02RE

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Stericup™ Quick Release-HV Sterile Vacuum Filtration System has a design and balanced profile that improves stability during filtration in tight laminar flow hoods.

The Stericup™ vacuum filtration system is the most efficient single-use, disposable process for sterile media preparation. It combines a Steritop™ filter unit with a receiver flask for processing and storing volumes from 150 mL to 1000 mL in a single, ready-to-use format.

Its no-tip, easy-grip flask design and balanced profile improve stability during filtration in tight laminar flow hoods, and the convenient labeling and packaging make it easy to open and track each device.

The bottom of the receiver flask is slightly recessed, so you can conveniently stack capped flasks after filtration, maximizing precious refrigeration space. You can also use Steritop™ bottle-top devices with your own sterile capture flask or bottle.

Features & Benefits

  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
  • Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
  • Stable base and easy grip design of the Stericup™ device enable any user to comfortably perform all stages of the workflow in a laminar flow hood.
  • Recessed bottom of Stericup receiver flasks allows flasks to be stacked easily in refrigerated spaces.

Applications

  • Sterilization of Aqueous Solutions (150 mL to 1 L)
    • Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.