Learn More
Kern Sohn Stereo Zoom Microscope OZP
Product Code. 17389541
Encompass_Preferred
Change view
Click to view available options
Head Style:
Binocular
Trinocular
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Head Style Unit Size
17389541 Trinocular Each
17379541 Binocular Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17389541 Supplier Kern Sohn Supplier No. OZP 558
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Professional and powerful thanks to its extremely large magnification range, strong illumination and first-class optics.
  • The KERN OZP stereo zoom microscope stands out through its above-average magnification range and its robust shape which is also ergonomic, it enables effortless, simple working over a period of several hours
  • The KERN OZP is available as a strong, continuously adjustable 3W LED reflected and transmitted light variant for the very best illumination of your sample
  • With its large working distance, an extra large field of view and its brilliant resolution, the KERN OZP provides sharp, high-contrast, colour-true images.
  • The extremely large, infinitely adjustable magnification range from 6 to 55 times magnification means that you can work quickly and effectively
  • The pillar stand is particularly flexible due to its variable and sturdy adjustment mechanism and therefore enables ergonomic working procedures.
  • A large selection of eyepieces, (universal) stands, a darkfield kit, external illumination units as well as auxiliary objectives and more are available as accessories
  • A protective dust cover, eye cups, as well as multi-lingual user instructions are included in the scope of delivery.
  • Scope of application: zoology and botany, quality control, Electronics and semiconductor industry, assembly and repair
  • Applications/Samples: Samples with focus on three-dimensional impression, zoom with variable magnification (depth, thickness), e.g. insects, seeds, circuit boards, components
Specifications
Voltage 230 V
Magnification Power 0.6X to 5.5X
Head Style Trinocular
Lighting Type LED