Learn More
Zeiss™ SteREO Discovery V8 with Fluorescence
Product Code. 17302840
Change view
Click to view available options
Includes:
LED double spot illuminator
Product Code. Includes
17302840 LED double spot illuminator
1 options
Product Code. 17302840 Supplier Zeiss™ Supplier No. 4950150019000
  • ONLINE EXCLUSIVE PRICE
    £14080.50
    £12954.00 / 1 sets
    Save £1126.50
    8% Off
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Stereomicroscope SteREO Discovery.V8 with fluorescence. Stand N with LED double spot illuminator. Intermediate LED tube 38HE for GFP.
  • The SteREO Discovery V8 offers crisp and brilliant images throughout the whole 8:1 manual zoom range.
  • The noticeably improved three dimensional image perception helps you to prepare and manipulate your samples - especially in developmental biology and quality assurance.
  • Acquire crisp and brilliant images throughout the whole 8:1 zoom range.
  • The selectable click-stops for discrete magnification steps allow you to calibrate pixel sizes in the imaging software packages ZEN and AxioVision.
Specifications
Includes LED double spot illuminator