missing translation for 'onlineSavingsMsg'
Learn More

StepOne™ StepOne™ Real-Time PCR System, Desktop

Product Code. 12170012 Shop All Applied Biosystems Products
Change view
Click to view available options
Content And Storage:
Desktop Computer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
12170012 Desktop Computer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12170012 Supplier StepOne™ Supplier No. 4376374
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Desktop Computer

StepOne™ Real-Time PCR System is a 48-well, low-throughput Real-Time PCR instrument perfect for both first-time and experienced users.

The StepOne™ Real-Time PCR System can be setup in a variety of configurations and comes ready to use, out of the box, with intuitive data analysis and instrument control software. Utilizing robust LED based 3-color optical recording, the StepOne™ Real-Time PCR System is designed to deliver precise, quantitative Real-Time PCR results for a variety of genomic research applications. Includes a tower computer.

Features of the The StepOne™ Real-Time PCR System include:
  • Advanced software and instrumentation for performing a wide array of genomic assays.
  • Sensitive 3-color optical LED recording syste
  • Intuitive and robust software perfect for both first-time and advanced users
  • Simple and flexible instrument set-up and usage
StepOne™ Software:
The StepOne™ Software included with the StepOne™ Real-Time PCR System runs on Windows™ XP, Window Vista™, and Windows™ 7 operating systems and provides instrument control, data collection, and data analysis capabilities. This latest version includes capabilities to collect melt curve data for High Resolution Melt (HRM) applications and the option to export in Real-Time PCR Data Markup Language (RDML) for compatibility with MIQE guidelines.

Rapid and Simple Assay Set-Up:
This remarkably simple Real-Time PCR system is designed with an LCD touchscreen and a user-friendly interface that brings the power of genetic studies to researchers new to Real-Time PCR. The intuitive software and protocol wizards help guide new users through their Real-Time PCR experiments. The StepOne™ Real-Time PCR System also includes a quick-start setup so that you start a run immediately and enter plate information at a later time.

Sensitive LED-Based 3-Color Fluorescence Reading:
The StepOne™ Real-Time PCR System utilizes a long-life LED-based optical system that can record fluorescence from FAM™/SYBR™ Green, VIC™/JOE™, and ROX™ dyes. This cost-effective, 3-color, 48-well system delivers precise, quantitative Real-Time PCR results and saves data from all filters in every run without depending on a computer or plate setup. It can discriminate between 2 populations of 5,000 and 10,000 template copies of a TaqMan™ assay with 99.7% confidence.

Compatible with Many Genomic Analysis Techniques:
Perform a variety of standard and demanding genetic analysis research techniques on one instrument using the user friendly StepOne™ Real-Time PCR System and software. The StepOne™ System supports many Real-Time PCR applications, including the following:
  • SNP Genotyping
  • Gene Expression Analysis
  • MicroRNA Expression
  • Translocation Analysis
  • Gene Detection
  • Viral Load Analysis
The included StepOne™ Software supports a variety of analysis methods, including the following:
  • Standard Curve (absolute quantitation)
  • Relative Standard Curve
  • Comparative Ct (ΔΔCt) (relative quantitation)
  • Genotyping and Presence/Absence
  • Melt Curve Analysis
  • High Resolution Melting (additional software required)

Versatile Instrument Configuration Options:
The ultra-compact footprint of a StepOne™ System can be installed in multiple distinct configurations, providing unmatched flexibility and convenience that can allow a fit in any laboratory. The StepOne™ Real-Time PCR System can be used via a PC, with a PC connected to a Local Area Network (LAN), or as a stand-alone instrument (PC-free). Each StepOne™ Real-Time PCR System is factory-calibrated for optical and thermal accuracy, so simply remarkable Real-Time PCR results can be obtained right out of the box.

Real-Time Data Monitoring, Dissemination, and Storage:
The system measures amplification as it occurs, allowing you to monitor the progress of your experiment cycle by cycle, either on the machine or remotely. Your data is stored on the instrument itself, and can be viewed and stored automatically via remote access or transferred via email or a USB flash drive. Data can also be conveniently exported as PowerPoint™, Excel™, and graphical image files.

Expanded Gene Expression Capabilities:
The advanced software provided with the StepOne™ Real-Time PCR System now includes the powerful Gene Expression Study Package. This software package allows for greater flexibility and accuracy in your gene expression assays through:
  • Analysis of an unlimited number of plates in one study
  • Sorting of data by biological or technical replicate group
  • Use of multiple endogenous controls
  • Assay efficiency correction

High Resolution Melting Capability:
Now with High Resolution Melt Software v3.0 you can perform post-PCR sequence variation analysis with the StepOne™ Real-Time PCR System. This separately purchased software simplifies set-up by maintaining assay specific settings and accepts pasted plate layout information directly from Excel. HRM Software v3.0 also has an improved clustering algorithm for increased sensitivity and accuracy, and the ability to conduct separate analyses of multiple assays run on a single plate.

For Research Use Only. Not for use in diagnostics procedures.
TRUSTED_SUSTAINABILITY

Specifications

Thermal Accuracy ± 0.25°C (35°C to 95°C) of Display Temperature
Sensitivity 1 Copy
Dynamic Range Linear Dynamic Range greater than 9 Log Units (Detection)
Content And Storage Desktop Computer
Thermal Uniformity ± 0.50°C (35°C to 95°C) of Setpoint/Display Temperature
Voltage 120 V
Height (Metric) 51.2 cm
Depth (Metric) 42.7 cm
Thermal Range 4°C to 100°C
For Use With (Application) Genotyping and Genomic Profiling, Real Time PCR (qPCR), Real Time PCR-based Gene Expression Profiling
Description Scan Time: <2 hr./run (Standard Mode), <40 min./run (Fast Mode)
Max. Heating Rate (Thermal Block) 4.6°C/s
Software Analysis Modes Fast mode: ±2.2°C/s, Standard mode: ±1.6°C/s
Volume (Metric) Thermal Block Sample 10 to 30μL (Standard Curve Experiments: 40μL in Standard Mode is Validated)
Multiplex Multiplexing
Detection 3 Emission Filters, LED, Photodiode
Quantity 1 Instrument
Pre-Calibrated Dyes FAM™, JOE™, ROX™, SYBR™ Green I, VIC™
Weight 24 kg (52 lb.)
Block Configurations 0.1 mL Tubes, 48-well Plate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.