missing translation for 'onlineSavingsMsg'
Learn More

STEP Antibody, DyLight 550, Novus Biologicals™

Product Code. 18410876 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18410876 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18410876 Supplier Novus Biologicals Supplier No. NBP281893R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STEP Polyclonal antibody specifically detects STEP in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen STEP
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias EC 3.1.3.48, FLJ14427, Neural-specific protein-tyrosine phosphatase, protein tyrosine phosphatase, non-receptor type 5 (striatum-enriched), protein-tyrosine phosphatase striatum-enriched, STEPPTPSTEP, Striatum-enriched protein-tyrosine phosphatase, tyrosine-protein phosphatase non-receptor type 5
Host Species Rabbit
Immunogen STEP antibody was raised against an 18 amino acid peptide near the amino terminus of human STEP. The immunogen is located within amino acids 180 - 230 of STEP. Amino Acid Squence: LRTPPEPPTPLPPEDRRQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Cellular Markers, Hypoxia, Neurodegeneration, Neuroscience, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84867
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.