missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ StemXVivo Serum-Free Human T Cell Base Media

Product Code. 18441170
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18441170 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18441170 Supplier R&D Systems™ Supplier No. CCM010
  • ONLINE EXCLUSIVE PRICE
    £247.79
    £218.40 / 250mL
    Save £29.39
    11% Off
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-optimized for ex vivo culture of human T lymphocytes

Isolation of human T-lymphocytes from peripheral blood is a time consuming process that can yield cells of varying purity and quality.

  • Supports T lymphocyte expansion better than RPMI containing 10% FBS
  • Decreases experimental variability through the use of defined media components
  • Can be used for multiple applications including mixed lymphocyte reactions (MLR), expansion of antigen-specific or non-specific T cells, and proliferation of peripheral blood lymphocytes in the presence of pharmacological agents such as PMA or ConA

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Media
Product Line R&D Systems
Quantity 250 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.