missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ StemXVivo Serum-Free Dendritic Cell Base Media

Product Code. 18412250
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18412250 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412250 Supplier R&D Systems™ Supplier No. CCM003
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-optimized base media for successful dendritic cell culture

The efficiency of differentiation of common myeloid progenitors into dendritic cells can vary from one experiment to another depending on the media components, the supplements used to drive differentiation, and the quality of the starting cell population.

  • High lot-to-lot consistency decreases variation
  • Can be supplemented with user-defined growth factors and cytokines
  • Optimized for differentiation of CD14+ monocytes into dendritic cells

TRUSTED_SUSTAINABILITY

Specifications

Product Type Stem Cell Media
Product Line R&D Systems
Quantity 250 mL
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.