missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies StemSpan™-ACF Erythroid Expansion Medium

Product Code. 17139961
Encompass
Change view
Click to view available options
Quantity:
500 mL Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17139961 500 mL Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17139961 Supplier STEMCELL Technologies Supplier No. 09860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Animal component-free medium for culture and expansion of human erythroid cells

  • StemSpan™-ACF Erythroid Expansion Medium (ACF-E) is an animal component-free (ACF) medium for the in vitro culture and expansion of human hematopoietic cells
  • When supplemented with StemSpan™ Erythroid Expansion Supplement (100X; Catalog #02692), this medium supports the expansion and erythroid differentiation of human hematopoietic progenitor cells into CD71+ and Glycophorin A+ erythroblasts in ACF culture conditions
  • This medium only contains recombinant proteins and synthetic components; it does not contain serum or other human- or animal-derived components

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells, Erythroid Cells
For Use With (Application) Cell Culture; Differentiation; Expansion
Quantity 500 mL Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.