missing translation for 'onlineSavingsMsg'
Learn More

StemRegenin 1, MedChemExpress

Product Code. 16486616
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16486616 100 mg 100mg
16462407 50 mg 50mg
16426288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16486616 Supplier Medchem Express Supplier No. HY15001
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress StemRegenin 1 is a potent aryl hydrocarbon receptor (AhR) antagonist with IC50 of 127 nM.

StemRegenin 1 is a potent aryl hydrocarbon receptor (AhR) antagonist with IC50 of 127 nM.

Specifications

Chemical Name or Material StemRegenin 1
CAS 1227633-49-9
Color White
Purity Grade Notes Research
Molecular Formula C24H23N5OS
Quantity 100 mg
Synonym SR1
Solubility Information DMSO : ≥ 100 mg/mL (232.81 mM)
SMILES CC(C)N1C2=NC(C3=CSC4=C3C=CC=C4)=NC(NCCC5=CC=C(O)C=C5)=C2N=C1
Molecular Weight (g/mol) 429.54
Formula Weight 429.54
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.