missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies StemRegenin 1
Description
- StemRegenin1 (SR1) is an antagonist of the aryl hydrocarbon receptor (AHR)
- It promotes ex vivo expansion of CD34+ human hematopoietic stem cells (Boitano et al.), 2010, Csazar et al.), 2012)
- SR1 has been shown to collaborate with UM729 in preventing differentiation of acute myeloid leukemia (AML) cells in culture (Pabst et al.), 2014)
- SR1 also stimulates the proliferation and differentiation of CD34+ hematopoietic progenitor cells into dendritic cells (Thordardottir et al.), 2014)
Specifications
Specifications
| CAS | 1227633-49-9 |
| Synonym | SR1 |
| Quantity | 5 mg |
| Molecular Formula | C24H23N5OS |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?