Learn More
Zeiss™ Stemi 305 Stereomicroscope with Flexible Stands
Product Code. 17352830
Change view
Click to view available options
Base Style:
Boom Stand B
Swiveling Stand U
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Base Style unitSize
17352830 Boom Stand B 1 set
17362830 Swiveling Stand U 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17352830 Supplier Zeiss™ Supplier No. 4919030010000
  • ONLINE EXCLUSIVE PRICE
    £3599.27
    £3056.00 / 1 sets
    Save £543.27
    15% Off
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket
Compact Greenough stereo microscope with 5:1 zoom for biological education, labs and production environments.
  • This compact Greenough stereo microscope allows you to observe your samples as they are: three-dimensional and crisp in contrast and no preparation required.
  • The Stemi 305 is an easy-to-use microscope, requires minimal setup, everything is integrated including long-living LED illumination for both reflected and transmitted light.
  • Spot Illuminator K LED - Single spot for brightfield and darkfield epi-illumination, Thread M24x0.5 for polarizer, Height adjustable, inclinable, zoomable.
Specifications
Base Style Boom Stand B
For Use With (Application) Biological Education, Labs and Production Environments