missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ T Cell Kit

Product Code. 17187491
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17187491 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17187491 Supplier STEMCELL Technologies Supplier No. 1000194
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For expansion and differentiation of hPSCs to T cells

  • STEMdiff™ T Cell Kit includes serum-free media and supplements for the feeder-free differentiation of human pluripotent stem cells (PSCs) to T cells
  • STEMdiff™ Hematopoietic - EB reagents are animal component-free and optimized for lymphoid differentiation potential
  • Using these reagents, CD34+ hematopoietic progenitor cells are first generated from PSCs; these can then be differentiated to T cells using the StemSpan™ components

Specifications

Cell Type Hematopoietic Cells, PSC-Derived, Pluripotent Stem Cells, T Cells, T Cells, CD8+
For Use With (Application) Cell Culture, Differentiation, Expansion
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.