missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ SMADi Neural Induction Kit, 2 Pack
Click to view available options
Quantity:
2 Kits
Unit Size:
1 set
Description
- STEMdiff™ SMADi Neural Induction Kit consists of a defined, serum-free medium and supplement for the highly efficient neural induction of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
- This kit combines STEMdiff™ Neural Induction Medium (Catalog #05835) with STEMdiff™ SMADi Neural Induction Supplement, which directs differentiation by blocking TGF-β/BMP-dependent SMAD signaling, resulting in efficient neural induction of even hard-to-differentiate cell lines
- Neural progenitor cells (NPCs) can be generated using STEMdiff™ SMADi Neural Induction Kit with either an embryoid body (EB) protocol or monolayer culture protocol
- The resulting cultures are enriched for central nervous system (CNS)-type NPCs, which express SOX1, Nestin, and PAX6
- NPCs generated using this kit can be passaged as single cells and expanded in STEMdiff™ Neural Progenitor Medium (Catalog #05833)
- The NPCs can also be differentiated into neurons and glia
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Pluripotent Stem Cells |
| For Use With (Application) | Cell Differentiation |
| Quantity | 2 Kits |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction