Learn More
STEMCELL Technologies STEMdiff™ Neural Rosette Selection Reagent
Description
- STEMdiff™ Neural Rosette Selection Reagent is an enzyme-free reagent for the selective detachment of neural rosette clusters from adherent neural aggregates previously generated from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells using STEMdiff™ Neural Induction Medium, without manual scraping
- Collecting and re-plating rosette clusters after incubation with STEMdiff™ Neural Rosette Selection Reagent will yield highly pure populations of neural progenitor cells, which can be further sub-cultured as single cells
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Pluripotent Stem Cells |
| Quantity | 100 mL |