missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Midbrain Neuron Differentiation Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
- STEMdiff™ Midbrain Neuron Differentiation Kit (Catalog #100-0038) is used to generate midbrain neuronal precursors from neural progenitor cells (NPCs) derived from human pluripotent stem cells (hPSCs) using STEMdiff™ SMADi Neural Induction Kit (Catalog #08581)
- The midbrain neuronal precursors are further matured into midbrain neurons using STEMdiff™ Midbrain Maturation Kit (Catalog #100-0041)
- These media will produce a population of midbrain neurons (≥ 15% TH-positive dopaminergic neurons; ≥ 90% class III βtubulin-positive neurons; < 10% GFAP-positive astrocytes)
- Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation
Specifications
Specifications
| Cell Type | Dopaminergic Neurons; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture; Differentiation |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction