missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Microglia Differentiation Kit
Description
- The STEMdiff™ Microglia Culture System comprises STEMdiff™ Microglia Differentiation Kit and STEMdiff™ Microglia Maturation Kit
- Together, these kits are used to differentiate and mature microglia derived from human pluripotent stem cells (hPSCs) using STEMdiff™ Hematopoietic Kit (Catalog #05310)
- Based on the protocol from the laboratory of Mathew Blurton-Jones (Abud et al.), 2017), the resulting cells are a highly pure population of microglia (> 80% CD45/CD11b-positive, > 50% TREM2-positive microglia; < 20% morphologically distinct monocytes or macrophages)
- Cells derived using these products are versatile tools for modeling neuroinflammation, studying human neurological development and disease, co-culture applications, and toxicity testing
Specifications
Specifications
| Cell Type | Hematopoietic Cells, PSC-Derived, Microglia, Neural Cells, PSC-Derived |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?