missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Kidney Organoid Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- STEMdiff™ Kidney Organoid Kit is a complete, serum-free cell culture medium system that supports highly efficient and reproducible generation of human pluripotent stem cell (hPSC)-derived kidney organoids in a simple, two-stage differentiation protocol
- These kidney organoids are composed of podocytes, proximal tubules, distal tubules, as well as the associated endothelium and mesenchyme
- Kidney organoids generated with STEMdiff™ Kidney Organoid Kit are tested for compatibility with phenotypic high-throughput assays such as nephrotoxic compound screening
- They also provide a relevant and convenient model for studies related to developmental biology and disease modeling
Specifications
Specifications
| Cell Type | Kidney Cells, Mesoderm, PSC-Derived |
| For Use With (Application) | Cell Culture, Differentiation, Functional Assay, Organoid Culture |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction