missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies STEMdiff™ Cardiomyocyte Dissociation Kit

Product Code. 16308405
Encompass
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16308405 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16308405 Supplier STEMCELL Technologies Supplier No. 5025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For dissociation of hPSC-derived cardiomyocytes

  • STEMdiff™ Cardiomyocyte Dissociation Kit includes STEMdiff™ Cardiomyocyte Dissociation Medium and STEMdiff™ Cardiomyocyte Support Medium
  • STEMdiff™ Cardiomyocyte Dissociation Medium can be used to harvest cardiomyocytes that have been differentiated from human pluripotent stem cells (hPSCs) using STEMdiff™ Cardiomyocyte Differentiation Kit (Catalog #05010) and maintained in STEMdiff™ Cardiomyocyte Maintenance Medium (Catalog #05010/05020)
  • STEMdiff™ Cardiomyocyte Support Medium reduces stress on these cardiomyocytes during harvesting and replating, maintaining their viability and functional capacity for downstream applications and analyses

Specifications

Cell Type Cardiomyocytes, PSC-Derived
For Use With (Application) Cell Culture
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.