missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies STEMdiff™ Astrocyte Maturation Kit
Description
- STEMdiff™ Astrocyte Maturation Kit is used to rapidly and efficiently generate cortical-type astrocytes from astrocytic precursors derived from human pluripotent stem cells (hPSCs) using the STEMdiff™ Astrocyte Differentiation Kit (Catalog #100-0016)
- Using this system, a highly pure population of astrocytes (> 70% S100B-positive and > 60% GFAP-positive astrocytes; < 15% doublecortin-positive neurons) can be generated from hPSCs in as few as 7 weeks and can be maintained long term in culture
- Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation
Specifications
Specifications
| Cell Type | Astrocytes, Neural Cells, PSC-Derived |
| For Use With (Application) | Cell Culture, Differentiation, Functional Assay |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?